ω-Bromo-2-chloroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α-Bromo-2-chloroacetophenone, Bromomethyl o-chlorophenyl ketone, 2-Bromo-1-(2-chlorophenyl)ethanone, 2-Bromo-2'-chloroacetophenone, 2-Chlorophenacyl bromide

017237-5G 017237-1G
FLUO017237-5GEA 0 GBP
FLUO017237-5G FLUO017237-1G
ω-Bromo-2-chloroacetophenone
ω-Bromo-2-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00832993
CAS Number: 5000-66-8
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR