1-Bromo-4-pentylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane

018302-1G 018302-5G
FLUO018302-1GEA 0 GBP
FLUO018302-1G FLUO018302-5G
1-Bromo-4-pentylbenzene
1-Bromo-4-pentylbenzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
MDL Number: MFCD00061113
CAS Number: 51554-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR