Phenylacetylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003009-25G 003009-100G 003009-500G
FLUO003009-25GEA 0 GBP
FLUO003009-25G FLUO003009-100G FLUO003009-500G
Phenylacetylene
Phenylacetylene
Formula: C₈H₆
MW: 102.14 g/mol
Boiling Pt: 144
Melting Pt: –44.8 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 31
Storage Temperature: Refrigerator
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3 III
Merck Index: 13,03892

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR