Thioanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
002945-500G
002945-100G
FLUO002945-500GEA
0
GBP
FLUO002945-500G
FLUO002945-100G
Thioanisole
Thioanisole
Formula:
C₇H₈S Boiling Pt: 188 °C (1013 hPa) Melting Pt: –15 °C Density: 1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008559 CAS Number: 100-68-5 EINECS: 202-878-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...