(Ethylthio)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Ethyl phenyl sulphide

001759-25G 001759-100G
FLUO001759-25GEA 0 GBP
FLUO001759-25G FLUO001759-100G
(Ethylthio)benzene
(Ethylthio)benzene
Formula: C₈H₁₀S
Boiling Pt: 37 °C (0.5 mmHg)
Melting Pt: 206.33 °C
Density: 1,0315 g/cm³ (10 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009265
CAS Number: 622-38-8
EINECS: 210-731-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR