2-Hydroxythiophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-Hydroxythiophenol, 2-Hydroxyphenyl mercaptan, 2-Hydroxybenzene-1-thiol, 1-Hydroxy-2-mercaptobenzene, 2-Mercaptophenol, 2-Hydroxybenzenethiol
M-Clarity™ quality level = MQ100
465348-1G
465348-5G
SIAL465348-1GEA
58.8
GBP
SIAL465348-1G
SIAL465348-5G
2-Hydroxythiophenol, Sigma-Aldrich®
2-Hydroxythiophenol
Formula:
C₆H₆OS Boiling Pt: 224 °C (1013 hPa) Melting Pt: 20 °C Density: 1.22 g/cm³ (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00040447 CAS Number: 1121-24-0 EINECS: 214-326-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...