2-Hydroxythiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-Hydroxythiophenol, 2-Hydroxyphenyl mercaptan, 2-Hydroxybenzene-1-thiol, 1-Hydroxy-2-mercaptobenzene, 2-Mercaptophenol, 2-Hydroxybenzenethiol
FL8733.1
008733-5G
008733-25G
CHMPFL8733.1EA
0
GBP
CHMPFL8733.1
FLUO008733-5G
FLUO008733-25G
2-Hydroxythiophenol
2-Hydroxythiophenol
Formula:
C₆H₆OS MW: 126.18 g/mol Boiling Pt: 224 Melting Pt: 20 Density: 1.22 g/cm³ (20 °C) Flash Pt: 97 |
MDL Number:
MFCD00040447 CAS Number: 1121-24-0 EINECS: 214-326-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...