Benzoic anhydride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
385980-100G
385980-500G
SIAL385980-100GEA
60.3
GBP
SIAL385980-100G
SIAL385980-500G
Benzoic anhydride, Sigma-Aldrich®
Benzoic anhydride
Formula:
C₁₄H₁₀O₃ MW: 226.23 g/mol Boiling Pt: 360 Melting Pt: 40 Density: 1.231…1.247 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003073 CAS Number: 93-97-0 EINECS: 202-291-1 Merck Index: 13,01093 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...