3,4-Dimethoxybenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005325-5G 005325-25G
FLUO005325-5GEA 0 GBP
FLUO005325-5G FLUO005325-25G
3,4-Dimethoxybenzenesulphonyl chloride
3,4-Dimethoxybenzenesulphonyl chloride
Formula: C₈H₉ClO₄S
Melting Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051769
CAS Number: 23095-31-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR