4-Methoxythiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: p-Methoxythiophenol, 4-Mercaptoanisol, 1-Methoxy-4-mercaptobenzene, 4-Methoxybenzenethiol, 4-Methoxybenzene-1-thiol, 4-Methoxyphenyl mercaptan

001774-25G 001774-10G
FLUO001774-25GEA 0 GBP
FLUO001774-25G FLUO001774-10G
4-Methoxythiophenol
4-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.1397 g/cm³ (20 °C)
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR