2-Methoxythiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003589-5G
003589-25G
003589-100G
FLUO003589-5GEA
0
GBP
FLUO003589-5G
FLUO003589-25G
FLUO003589-100G
2-Methoxythiophenol
2-Methoxythiophenol
Formula:
C₇H₈OS Boiling Pt: 225 °C (1013 hPa) Density: 1.14 g/cm³ (30 °C) |
MDL Number:
MFCD00004834 CAS Number: 7217-59-6 EINECS: 230-605-7 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...