2-Nitroresorcinol 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Nitrobenzene-1,3-diol

416160100 416160250
ACRO416160100EA 60.2 GBP
ACRO416160100 ACRO416160250
2-Nitroresorcinol 98%
2-Nitroresorcinol
Formula: C₆H₅NO₄
MW: 155.11 g/mol
Boiling Pt: 232
Melting Pt: 80…84 °C
MDL Number: MFCD00007124
CAS Number: 601-89-8
EINECS: 210-010-9

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
Water ≤2 %
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR