3-Nitrophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
m-Nitrophenol
M-Clarity™ quality level = MQ100
163031-10G
163031-50G
SIAL163031-10GEA
61.9
GBP
SIAL163031-10G
SIAL163031-50G
3-Nitrophenol, Sigma-Aldrich®
3-Nitrophenol
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 271 °C (1013 hPa) Melting Pt: 94…95 °C Density: 1.485 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007240 CAS Number: 554-84-7 EINECS: 209-073-5 UN: 1663 ADR: 6.1,III Merck Index: 13,06652 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...