2-Nitrophenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Nitrophenol

M-Clarity™ quality level = MQ200

N19702-100G N19702-1KG N19702-500G N19702-5G
SIALN19702-100GEA 65 GBP
SIALN19702-100G SIALN19702-1KG SIALN19702-500G SIALN19702-5G
2-Nitrophenol, Sigma-Aldrich®
2-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 214.6 °C (1013 hPa)
Melting Pt: 43…45 °C
Density: 1.599 g/cm³ (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1,III
Merck Index: 12,06717

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR