Nitrobenzene
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
4394.1
4394.2
ROTH4394.1EA
55.8
GBP
ROTH4394.1
ROTH4394.2
Nitrobenzene
Nitrobenzene
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Boiling Pt: 210.8 °C (1013 hPa) Melting Pt: 5.7 °C Density: 1.2037 g/cm³ (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007043 CAS Number: 98-95-3 EINECS: 202-716-0 UN: 1662 ADR: 6.1,II Merck Index: 13,06621 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...