p-Phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine
78429-100G
78429-250G
SIAL78429-100GEA
228
GBP
SIAL78429-100G
SIAL78429-250G
p-Phenylenediamine, Sigma-Aldrich®
p-Phenylenediamine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 267 °C (1013 hPa) Melting Pt: 139…141 °C Density: 1.251 g/cm³ (20 °C) Flash Pt: 156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007901 CAS Number: 106-50-3 EINECS: 203-404-7 UN: 1673 ADR: 6.1,III Merck Index: 12,07439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...