2-Aminothiophenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

274240-100G 274240-25G
SIAL274240-100GEA 113 GBP
SIAL274240-100G SIAL274240-25G
2-Aminothiophenol, Sigma-Aldrich®
2-Aminothiophenol
Formula: C₆H₇NS
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR