2-Aminothiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002470-100G 002470-500G
FLUO002470-100GEA 0 GBP
FLUO002470-100G FLUO002470-500G
2-Aminothiophenol
2-Aminothiophenol
Formula: C₆H₇NS
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR