3,4,5-Trimethoxyaniline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
T68209-10G
T68209-25G
SIALT68209-10GEA
64.9
GBP
SIALT68209-10G
SIALT68209-25G
3,4,5-Trimethoxyaniline, Sigma-Aldrich®
3,4,5-Trimethoxyaniline
Formula:
C₉H₁₃NO₃ MW: 183.21 g/mol Boiling Pt: 293 °C (1013 hPa) Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008393 CAS Number: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...