2-Amino-3-nitrophenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxy-6-nitroaniline

M-Clarity™ quality level = MQ100

297003-25G 297003-5G
SIAL297003-25GEA 92.9 GBP
SIAL297003-25G SIAL297003-5G
2-Amino-3-nitrophenol, Sigma-Aldrich®
2-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00010875
CAS Number: 603-85-0
EINECS: 210-060-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR