3,4,5-Trimethoxyphenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Isothiocyanato-3,4,5-trimethoxybenzene, 5-Isothiocyanato-1,2,3-trimethoxybenzene
009584-1G
009584-5G
FLUO009584-1GEA
0
GBP
FLUO009584-1G
FLUO009584-5G
3,4,5-Trimethoxyphenyl isothiocyanate
3,4,5-Trimethoxyphenyl isothiocyanate
Formula:
C₁₀H₁₁NO₃S MW: 225.27 g/mol Boiling Pt: 323 °C (1013 hPa) Melting Pt: 63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041076 CAS Number: 35967-24-9 EINECS: 252-820-5 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...