4-Methoxyphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxyphenylisothiocyanate

001278-25G 001278-5G
FLUO001278-25GEA 0 GBP
FLUO001278-25G FLUO001278-5G
4-Methoxyphenyl isothiocyanate
4-Methoxyphenyl isothiocyanate
Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 279…281 °C (1013 hPa)
Melting Pt: 17…19 °C
Density: 1,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011676
CAS Number: 2284-20-0
EINECS: 218-921-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR