3-Methoxyphenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methoxyphenylisothiocyanate
009513-25G
009513-5G
FLUO009513-25GEA
0
GBP
FLUO009513-25G
FLUO009513-5G
3-Methoxyphenyl isothiocyanate
3-Methoxyphenyl isothiocyanate
Formula:
C₈H₇NOS MW: 165.22 g/mol Boiling Pt: 84 °C (1 mmHg) Density: 1,1788 g/cm³ (20 °C) |
MDL Number:
MFCD00040335 CAS Number: 3125-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...