2-Methoxyphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001276-5G 001276-25G
FLUO001276-5GEA 0 GBP
FLUO001276-5G FLUO001276-25G
2-Methoxyphenyl isothiocyanate
2-Methoxyphenyl isothiocyanate
Formula: C₈H₇NOS
MW: 165.22 g/mol
MDL Number: MFCD00011675
CAS Number: 3288-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR