2-Ethoxyphenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Ethoxyphenylisothiocyanate
008941-5G
008941-25G
FLUO008941-5GEA
0
GBP
FLUO008941-5G
FLUO008941-25G
2-Ethoxyphenyl isothiocyanate
2-Ethoxyphenyl isothiocyanate
Formula:
C₉H₉NOS MW: 179.24 g/mol |
MDL Number:
MFCD00046793 CAS Number: 23163-84-0 EINECS: 245-468-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...