2-Methoxy-5-nitrophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009511-1G 009511-5G
FLUO009511-1GEA 0 GBP
FLUO009511-1G FLUO009511-5G
2-Methoxy-5-nitrophenyl isothiocyanate
2-Methoxy-5-nitrophenyl isothiocyanate
Formula: C₈H₆N₂O₃S
MW: 210.21 g/mol
MDL Number: MFCD00060634
CAS Number: 71793-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR