4-Nitrophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002722-5G
002722-25G
FLUO002722-5GEA
0
GBP
FLUO002722-5G
FLUO002722-25G
4-Nitrophenyl isothiocyanate
4-Nitrophenyl isothiocyanate
Formula:
C₇H₄N₂O₂S Boiling Pt: 331 °C (1013 hPa) Melting Pt: 108…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007307 CAS Number: 2131-61-5 EINECS: 218-359-9 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...