Phenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
78780-100ML
78780-25ML
78780-500ML
SIAL78780-100MLEA
166
GBP
SIAL78780-100ML
SIAL78780-25ML
SIAL78780-500ML
Phenyl isothiocyanate, Sigma-Aldrich®
Phenyl isothiocyanate
Formula:
C₇H₅NS MW: 135.19 g/mol Boiling Pt: 221 °C (1013 hPa) Melting Pt: –21 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004798 CAS Number: 103-72-0 EINECS: 203-138-1 UN: 2927 ADR: 6.1,II Merck Index: 12,07450 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...