3-Iodoaniline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
I7209-25G
I7209-5G
SIALI7209-25GEA
92.8
GBP
SIALI7209-25G
SIALI7209-5G
3-Iodoaniline, Sigma-Aldrich®
Formula:
C₆H₆IN Boiling Pt: 271 °C (1013 hPa) Melting Pt: 24…26 °C Density: 1.821 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007781 CAS Number: 626-01-7 EINECS: 210-922-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...