4-Iodophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008984-5G 008984-50G
FLUO008984-5GEA 0 GBP
FLUO008984-5G FLUO008984-50G
4-Iodophenyl isothiocyanate
4-Iodophenyl isothiocyanate
Formula: C₇H₄INS
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 75…78 °C
MDL Number: MFCD00041091
CAS Number: 2059-76-9
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR