2-Iodophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Iodo-2-isothiocyanatobenzene

008982-5G 008982-25G
FLUO008982-5GEA 0 GBP
FLUO008982-5G FLUO008982-25G
2-Iodophenyl isothiocyanate
2-Iodophenyl isothiocyanate
Formula: C₇H₄INS
MW: 261.09 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 39 °C
MDL Number: MFCD00041057
CAS Number: 98041-44-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR