3-Iodobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Iodobenzoic acid

M-Clarity™ quality level = MQ200

138584-10G 138584-50G
SIAL138584-10GEA 62.6 GBP
SIAL138584-10G SIAL138584-50G
3-Iodobenzoic acid, Sigma-Aldrich®
3-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 186…188 °C
Density: 2.171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9
EINECS: 210-555-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR