Methyl-3-iodobenzoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

706388-25G 706388-5G
SIAL706388-25GEA 226 GBP
SIAL706388-25G SIAL706388-5G
Methyl-3-iodobenzoate, Sigma-Aldrich®
Methyl 3-iodobenzoate
Formula: C₈H₇IO₂
MDL Number: MFCD00061093
CAS Number: 618-91-7
EINECS: 210-570-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR