Methyl-2-iodobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005544-25G 005544-100G
FLUO005544-25GEA 0 GBP
FLUO005544-25G FLUO005544-100G
Methyl-2-iodobenzoate
Methyl 2-iodobenzoate
Formula: C₈H₇IO₂
Boiling Pt: 272 °C (1013 hPa)
Density: 1,784 g/cm³ (20 °C)
MDL Number: MFCD00016351
CAS Number: 610-97-9
EINECS: 210-243-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR