Iodobenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Phenyliodide

M-Clarity™ quality level = MQ200

I7632-100G-A I7632-500G-A I7632-5G-A
SIALI7632-100G-AEA 75.9 GBP
SIALI7632-100G-A SIALI7632-500G-A SIALI7632-5G-A
Iodobenzene, Sigma-Aldrich®
Iodobenzene
Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.838 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR