Pentafluorobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006949-5G 006949-25G
FLUO006949-5GEA 0 GBP
FLUO006949-5G FLUO006949-25G
Pentafluorobenzenesulphonyl chloride
Pentafluorobenzenesulphonyl chloride
Formula: C₆ClF₅O₂S
Boiling Pt: 210 °C (1013 hPa)
Density: 1.79 g/cm³ (20 °C)
MDL Number: MFCD00007427
CAS Number: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR