Pentafluorothiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3,4,5,6-Pentafluorothiophenol, Pentafluorophenyl mercaptan, Pentafluorobenzenethiol, Mercaptopentafluorobenzene

003278-5G 003278-25G 003278-100G
FLUO003278-5GEA 0 GBP
FLUO003278-5G FLUO003278-25G FLUO003278-100G
Pentafluorothiophenol
Pentafluorothiophenol
Formula: C₆HF₅S
MW: 200.13 g/mol
Boiling Pt: 143
Melting Pt: –24 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004828
CAS Number: 771-62-0
EINECS: 212-236-3
UN: 2920

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR