4-Fluorophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
M-Clarity™ quality level = MQ100
F13207-100G
F13207-25G
SIALF13207-100GEA
85
GBP
SIALF13207-100G
SIALF13207-25G
4-Fluorophenol, Sigma-Aldrich®
4-Fluorophenol
Formula:
C₆H₅FO MW: 112.1 g/mol Boiling Pt: 185 °C (1013 hPa) Melting Pt: 44…47 °C Density: 1.22 g/cm³ (20 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002316 CAS Number: 371-41-5 EINECS: 206-736-0 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...