3-Fluorophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
F13002-100G
F13002-10G
F13002-50G
SIALF13002-100GEA
260
GBP
SIALF13002-100G
SIALF13002-10G
SIALF13002-50G
3-Fluorophenol, Sigma-Aldrich®
3-Fluorophenol
Formula:
C₆H₅FO MW: 112.1 g/mol Boiling Pt: 178 °C (1013 hPa) Melting Pt: 13.7 °C Density: 1.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002254 CAS Number: 372-20-3 EINECS: 206-748-6 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...