3-(Trifluoromethoxy)thiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trifluoromethoxy)benzenethiol, 3-(Trifluoromethoxy)benzene-1-thiol

013053-1G 013053-5G
FLUO013053-1GEA 0 GBP
FLUO013053-1G FLUO013053-5G
3-(Trifluoromethoxy)thiophenol
3-(Trifluoromethoxy)thiophenol
Formula: C₇H₅F₃OS
MW: 194.18 g/mol
MDL Number: MFCD01320799
CAS Number: 220239-66-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR