4-(Trifluoromethoxy)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α,α,α-Trifluoro-4-hydroxyanisole

001424-5G 001424-100G 001424-25G
FLUO001424-5GEA 0 GBP
FLUO001424-5G FLUO001424-100G FLUO001424-25G
4-(Trifluoromethoxy)phenol
4-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 17…18 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040988
CAS Number: 828-27-3
EINECS: 212-583-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR