2-Fluoro-4-nitroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

006292-1G 006292-5G 006292-25G
FLUO006292-1GEA 0 GBP
FLUO006292-1G FLUO006292-5G FLUO006292-25G
2-Fluoro-4-nitroanisole
2-Fluoro-4-nitroanisole
Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD00061095
CAS Number: 455-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR