1-(Difluoromethoxy)-2-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004839-1G 004839-5G
FLUO004839-1GEA 0 GBP
FLUO004839-1G FLUO004839-5G
1-(Difluoromethoxy)-2-nitrobenzene
1-(Difluoromethoxy)-2-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
MDL Number: MFCD00221470
CAS Number: 22225-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR