1-Fluoro-2-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
o-Fluoronitrobenzene, 2-Fluoronitrobenzene
F10801-250G
F10801-50G
SIALF10801-250GEA
126
GBP
SIALF10801-250G
SIALF10801-50G
1-Fluoro-2-nitrobenzene, Sigma-Aldrich®
1-Fluoro-2-nitrobenzene
Formula:
C₆H₄FNO₂ Boiling Pt: 212 °C (1013 hPa) Melting Pt: –8 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007048 CAS Number: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...