2-Amino-5-fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013054-1G 013054-5G
FLUO013054-1GEA 0 GBP
FLUO013054-1G FLUO013054-5G
2-Amino-5-fluorophenol
2-Amino-5-fluorophenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
MDL Number: MFCD00671759
CAS Number: 53981-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR