4-(Trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001400-25G 001400-100G 001400-500G
FLUO001400-25GEA 0 GBP
FLUO001400-25G FLUO001400-100G FLUO001400-500G
4-(Trifluoromethoxy)aniline
4-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
Boiling Pt: 73 °C (1013 hPa)
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR