4-(Difluoromethoxy)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004828-5G
004828-25G
FLUO004828-5GEA
0
GBP
FLUO004828-5G
FLUO004828-25G
4-(Difluoromethoxy)aniline
4-(Difluoromethoxy)aniline
Formula:
C₇H₇F₂NO MW: 159.14 g/mol Boiling Pt: 90 Density: 1.286 g/cm³ (20 °C) |
MDL Number:
MFCD00085005 CAS Number: 22236-10-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...