3-(Trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001474-5G 001474-25G
FLUO001474-5GEA 0 GBP
FLUO001474-5G FLUO001474-25G
3-(Trifluoromethoxy)aniline
3-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
Boiling Pt: 72 °C (1013 hPa)
Density: 1.3254 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041511
CAS Number: 1535-73-5
EINECS: 216-256-3
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR