2-(Trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001409.1G 001409-5G 001409-100G 001409-25G
FLUO001409.1GEA 0 GBP
FLUO001409.1G FLUO001409-5G FLUO001409-100G FLUO001409-25G
2-(Trifluoromethoxy)aniline
2-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
Boiling Pt: 64.5 °C (21 mmHg)
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00035959
CAS Number: 1535-75-7
EINECS: 216-257-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR