2-(Difluoromethoxy)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
007849-5G
007849-25G
FLUO007849-5GEA
0
GBP
FLUO007849-5G
FLUO007849-25G
2-(Difluoromethoxy)aniline
2-(Difluoromethoxy)aniline
Formula:
C₇H₇F₂NO Boiling Pt: 86 °C (12 mmHg) Density: 1.265 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00190104 CAS Number: 22236-04-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...